missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-chloro-3-pyridyl)pyrimidine, 97%, Thermo Scientific™

Product Code. 15411098
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as primary and secondary intermediate.

Notes
Incompatible with strong oxidizing agents, bases, acids. Keep in room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870221-49-1
Molecular Formula C9H7ClN4
Molecular Weight (g/mol) 206.633
MDL Number MFCD18157687
InChI Key DKCVVHMYFZBDAK-UHFFFAOYSA-N
PubChem CID 53396335
IUPAC Name 4-(2-chloropyridin-3-yl)pyrimidin-2-amine
SMILES C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N

Specifications

Quantity 250 mg
Beilstein 14484335
Synonym 4-2-chloropyridin-3-yl pyrimidin-2-amine, chloropyridinylpyrimidinamine, 2-pyrimidinamine,4-2-chloro-3-pyridinyl, 2-pyrimidinamine, 4-2-chloro-3-pyridinyl
Formula Weight 206.63
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(2-chloro-3-pyridyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'