missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-3-methylbenzoic acid, 97%, Thermo Scientific™ Product Code.: 15485217

2-Bromo-3-methylbenzoic acid, 97%, Thermo Scientific™

Product Code. 15485217
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15485217

Brand: Thermo Scientific Alfa Aesar H32490.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53663-39-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00079721
InChI Key LSRTWJCYIWGKCQ-UHFFFAOYSA-N
Synonym 2-bromo-3-methyl-benzoic acid, 2-bromo-m-toluic acid, 2-bromo-3-carboxytoluene, 2-bromo-3-methylbenzoicacid, 3-methyl-2-bromobenzoic acid, benzoic acid, bromo-3-methyl, 2-brom-3-methylbenzoic acid, pubchem3805, acmc-1ay3t, ksc497o4r
PubChem CID 2735588
IUPAC Name 2-bromo-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1Br)C(=O)O

Specifications

Melting Point 134°C to 137°C
Quantity 1 g
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-methylbenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10196151 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10175066 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10162858 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-3-methylbenzoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.