missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-4,5-dichloroimidazole, 98%, Thermo Scientific Chemicals Product Code.: 11472697

2-Bromo-4,5-dichloroimidazole, 98%, Thermo Scientific Chemicals

Product Code. 11472697
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11472697

Brand: Thermo Scientific Alfa Aesar A14896.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16076-27-0
Molecular Formula C3HBrCl2N2
Molecular Weight (g/mol) 215.86
MDL Number MFCD00067894
InChI Key SULQOTGFPPFUMG-UHFFFAOYSA-N
Synonym 2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole
PubChem CID 99484
IUPAC Name 2-bromo-4,5-dichloro-1H-imidazole
SMILES ClC1=C(Cl)N=C(Br)N1

Specifications

Melting Point ∼215°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 606732
Formula Weight 215.87
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,5-dichloroimidazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246031 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA14896.03,A14896.06
Lot Number10246032 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA14896.03,A14896.06
Lot Number10238348 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14896.03,A14896.06
Lot Number10238454 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14896.03,A14896.06
Lot NumberUSLF003542 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14896.03,A14896.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-4,5-dichloroimidazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.