missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chlorotoluene, 98%, Thermo Scientific Chemicals

Product Code. 11481000
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27139-97-5
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00060650
InChI Key CSUUXPHPCXHYGY-UHFFFAOYSA-N
Synonym 2-bromo-4-chlorotoluene, benzene, 2-bromo-4-chloro-1-methyl, 4-chloro-2-bromotoluene, toluene, 2-bromo-4-chloro, p-chlor brom toluol, pubchem3599, acmc-209gvg, 2-bromo-4-chloro toluene, 2-bromo-4-chloro-toluene, ksc493k9b
PubChem CID 609898
IUPAC Name 2-bromo-4-chloro-1-methylbenzene
SMILES CC1=C(Br)C=C(Cl)C=C1

Specifications

Quantity 25 g
Formula Weight 205.48
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chlorotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'