missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-fluorobenzyl alcohol, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Bromo-4-fluorobenzyl alcohol, is used as an alcohol, medicine intermediate.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Bromo-4-fluorobenzyl alcohol, is used as an alcohol, medicine intermediate.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | 68°C to 72°C |
Color | White |
Quantity | 25 g |
Synonym | 2-bromo-4-fluorophenyl methanol, 2-bromo-4-fluorobenzyl alcohol, benzenemethanol, 2-bromo-4-fluoro, 2-bromo-4-fluorophenyl methan-1-ol, 2-bromo-4-fluorobenzenemethanol, benzenemethanol,2-bromo-4-fluoro, pubchem3222, acmc-1cqar, ksc550i1h, rarechem al bd 1088 |
Solubility Information | Soluble in water. |
Formula Weight | 205.02 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 2-Bromo-4-fluorobenzyl alcohol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Bromo-4-fluorobenzyl alcohol, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction