missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromoacetophenone, 99%, Thermo Scientific Chemicals

694.00 SEK - 9660.00 SEK

Chemical Identifiers

CAS 2142-69-0
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000067
InChI Key PIMNFNXBTGPCIL-UHFFFAOYSA-N
Synonym 2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketoneShow MoreShow Less
PubChem CID 75060
IUPAC Name 1-(2-bromophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11464014
SDS View Documents
Thermo Scientific Alfa Aesar
A11982.09
10 g
694.00 SEK
10g
Please to purchase this item. Need a web account? Register with us today!
11474014
SDS View Documents
Thermo Scientific Alfa Aesar
A11982.18
50 g
2445.00 SEK
50g
Please to purchase this item. Need a web account? Register with us today!
11484014
SDS View Documents
Thermo Scientific Alfa Aesar
A11982.30
250 g
9660.00 SEK
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 218-398-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2'-Bromoacetophenone, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.