missing translation for 'onlineSavingsMsg'
Learn More

2-Butynal diethyl acetal, 98%, Thermo Scientific Chemicals

Product Code. 11327625
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Butynal diethyl acetal is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butynal diethyl acetal is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2806-97-5
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00009274
InChI Key FKKVKKSEVMQYER-UHFFFAOYSA-N
Synonym 2-butynal diethyl acetal, 2-butyn-1-al diethyl acetal, 2-butynyl aldehyde diethyl acetal, 2-butyne, 1,1-diethoxy, 1,1-diethoxy-but-2-yne, 1-diethoxybut-2-yne, acmc-20ao12, tetrolaldehyde diethyl acetal, 1,1-diethoxy-2-butyne #
PubChem CID 137721
IUPAC Name 1,1-diethoxybut-2-yne
SMILES CCOC(OCC)C#CC

Specifications

Density 0.898
Boiling Point 65°C to 67°C (12 mmHg)
Flash Point 56°C (133°F)
Odor Recognizable
Refractive Index 1.426
Quantity 1 g
UN Number UN1993
Beilstein 1702075
Solubility Information Not miscible in water.
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material 2-Butynal diethyl acetal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Butynal diethyl acetal, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.