missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-fluoro-4-iodobenzene, 98%, Thermo Scientific Chemicals

1315.00 SEK - 4240.00 SEK

Chemical Identifiers

CAS 156150-67-3
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.443
MDL Number MFCD00042210
InChI Key OMASDGWBVAVFQZ-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoroiodobenzene, 1-chloro-2-fluoro-5-iodobenzene, 2-chloro4-iodofluorobenzene, 3-chloro-4-fluoro-1-iodobenzene, 1-fluoro-2-chloro-4-iodobenzene, benzene, 2-chloro-1-fluoro-4-iodo, benzene,2-chloro-1-fluoro-4-iodo, 2-chloro-1-fluoro-4-iodo-benzene, pubchem3433, acmc-20an6s
PubChem CID 2724605
IUPAC Name 2-chloro-1-fluoro-4-iodobenzene
SMILES C1=CC(=C(C=C1I)Cl)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11419403
SDS View Documents
Thermo Scientific Alfa Aesar
A11808.14
25 g
1315.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
11429403
SDS View Documents
Thermo Scientific Alfa Aesar
A11808.22
100 g
4240.00 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

156150-67-3
256.443
OMASDGWBVAVFQZ-UHFFFAOYSA-N
2724605
C1=CC(=C(C=C1I)Cl)F
C6H3ClFI
MFCD00042210
3-chloro-4-fluoroiodobenzene, 1-chloro-2-fluoro-5-iodobenzene, 2-chloro4-iodofluorobenzene, 3-chloro-4-fluoro-1-iodobenzene, 1-fluoro-2-chloro-4-iodobenzene, benzene, 2-chloro-1-fluoro-4-iodo, benzene,2-chloro-1-fluoro-4-iodo, 2-chloro-1-fluoro-4-iodo-benzene, pubchem3433, acmc-20an6s
2-chloro-1-fluoro-4-iodobenzene

Specifications

156150-67-3
94°C to 95°C (15 mmHg)
C6H3ClFI
MFCD00042210
Slightly soluble in water.
C1=CC(=C(C=C1I)Cl)F
256.443
256.45
2-Chloro-1-fluoro-4-iodobenzene
2.02
>110°C (230°F)
1.605
3-chloro-4-fluoroiodobenzene, 1-chloro-2-fluoro-5-iodobenzene, 2-chloro4-iodofluorobenzene, 3-chloro-4-fluoro-1-iodobenzene, 1-fluoro-2-chloro-4-iodobenzene, benzene, 2-chloro-1-fluoro-4-iodo, benzene,2-chloro-1-fluoro-4-iodo, 2-chloro-1-fluoro-4-iodo-benzene, pubchem3433, acmc-20an6s
OMASDGWBVAVFQZ-UHFFFAOYSA-N
2-chloro-1-fluoro-4-iodobenzene
2724605
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-1-fluoro-4-iodobenzene, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.