missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-3,6-difluorobenzaldehyde, 97%, Thermo Scientific™ Product Code.: 11406716

2-Chloro-3,6-difluorobenzaldehyde, 97%, Thermo Scientific™

Product Code. 11406716
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11406716

Brand: Thermo Scientific Alfa Aesar B23866.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261762-39-4
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD01631443
InChI Key KPYBGKSDBNMHTO-UHFFFAOYSA-N
Synonym benzaldehyde, 2-chloro-3,6-difluoro, 2,5-difluoro-6-chlorobenzaldehyde, 6-chloro-2,5-difluoro-benzaldehyde, pubchem2618, zlchem 1271, acmc-1cfb6, intermediates-zcf02699, ksc495g2n, 2-chloro-3,6-difluoro-benzaldehyde, 2-chloro-3,6-difluorobenzaldehyde
PubChem CID 2773509
IUPAC Name 2-chloro-3,6-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1F)C=O)Cl)F

Specifications

Melting Point 46°C to 48°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 176.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-3,6-difluorobenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-3,6-difluorobenzaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.