missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-5-nitrobenzoic Acid, 99+%, Thermo Scientific Chemicals Product Code.: 10377420

2-Chloro-5-nitrobenzoic Acid, 99+%, Thermo Scientific Chemicals

Product Code. 10377420
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 10377420

Brand: Thermo Scientific Acros 154695000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-96-3
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.56
MDL Number MFCD00007294
InChI Key QUEKGYQTRJVEQC-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzoic acid, benzoic acid, 2-chloro-5-nitro, 6-chloro-3-nitrobenzoic acid, 2-chloro-5-nitro-benzoic acid, 2-chloro-5-nitrobenzoicacid, mesalamine impurity m, pubchem13682, acmc-1cb0i, 2-chloro-5-nitrobenzic acid, 5-nitro-2-chlorobenzoic acid
PubChem CID 17287
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 164°C to 168°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Plastic bottle
Linear Formula ClC6H3(NO2)CO2H
Quantity 500 g
Beilstein 09, 403
Solubility Information Solubility in water: insoluble
Formula Weight 201.56
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzoic acid, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0396190 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number154695000,154691000
Lot NumberA0413157 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number154695000,154691000
Lot NumberA0438606 Certificate TypeCertificate of Analysis Date03/02/2022 Catalog Number154695000,154691000
compliance-icons
Product Identifier
  • 2-Chloro-5-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-5-nitrobenzoic Acid, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.