Learn More
2-Chlorobenzoxazole, 98%, Thermo Scientific Chemicals
629.00 SEK - 1880.00 SEK
Chemical Identifiers
CAS | 615-18-9 |
---|---|
Molecular Formula | C7H4ClNO |
Molecular Weight (g/mol) | 153.57 |
MDL Number | MFCD00005766 |
InChI Key | BBVQDWDBTWSGHQ-UHFFFAOYSA-N |
Synonym | 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazoleShow More |
PubChem CID | 11986 |
IUPAC Name | 2-chloro-1,3-benzoxazole |
SMILES | ClC1=NC2=CC=CC=C2O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302137
|
Thermo Scientific Alfa Aesar
L07981.06 |
5 g |
629.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312137
|
Thermo Scientific Alfa Aesar
L07981.14 |
25 g |
1880.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Not miscible in water.
Notes
Store under inert gas. Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Keep away from heat.
Chemical Identifiers
615-18-9 | |
153.57 | |
BBVQDWDBTWSGHQ-UHFFFAOYSA-N | |
11986 | |
ClC1=NC2=CC=CC=C2O1 |
C7H4ClNO | |
MFCD00005766 | |
2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazoleShow More | |
2-chloro-1,3-benzoxazole |
Specifications
615-18-9 | |
201°C to 202°C | |
C7H4ClNO | |
MFCD00005766 | |
2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole | |
BBVQDWDBTWSGHQ-UHFFFAOYSA-N | |
2-chloro-1,3-benzoxazole | |
11986 | |
99% |
1.39 | |
85°C (185°F) | |
1.567 | |
115982 | |
Not miscible in water. | |
ClC1=NC2=CC=CC=C2O1 | |
153.57 | |
153.57 | |
2-Chlorobenzoxazole |
Safety and Handling
GHS H Statement
H302-H315-H319-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
H227-H302-H315-H319-H335
missing translation for 'einecsNumber' : 210-414-5
missing translation for 'rtecsNumber' : DM4680000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold