Learn More
(2-Chloroethyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
463.05 SEK - 1300.20 SEK
Chemical Identifiers
CAS | 999-81-5 |
---|---|
Molecular Formula | C5H13Cl2N |
Molecular Weight (g/mol) | 158.07 |
MDL Number | MFCD00011869 |
InChI Key | UHZZMRAGKVHANO-UHFFFAOYSA-M |
Synonym | chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan |
PubChem CID | 13836 |
IUPAC Name | 2-chloroethyl(trimethyl)azanium;chloride |
SMILES | C[N+](C)(C)CCCl.[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10052680
|
Thermo Scientific Acros
215420250 |
25 g |
463.05 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10246890
|
Thermo Scientific Acros
215421000 |
100 g |
1300.20 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
999-81-5 | |
158.07 | |
UHZZMRAGKVHANO-UHFFFAOYSA-M | |
13836 | |
C[N+](C)(C)CCCl.[Cl-] |
C5H13Cl2N | |
MFCD00011869 | |
chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan | |
2-chloroethyl(trimethyl)azanium;chloride |
Specifications
999-81-5 | |
100.0 | |
White | |
98% | |
C5H13Cl2N | |
MFCD00011869 | |
15, 2104 | |
Solubility in water: 740g/L (20°C) | |
C[N+](C)(C)CCCl.[Cl-] | |
158.07 | |
158.07 | |
Crystalline Powder or Crystals |
97.5 | |
239°C to 243°C | |
Authentic | |
Glass bottle | |
ClCH2CH2N(CH3)3Cl | |
04, 134 | |
chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan | |
UHZZMRAGKVHANO-UHFFFAOYSA-M | |
2-chloroethyl(trimethyl)azanium;chloride | |
13836 | |
98% | |
(2-Chloroethyl)trimethylammonium chloride, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-666-4
missing translation for 'rtecsNumber' : BP5250000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.