missing translation for 'onlineSavingsMsg'
Learn More

(2-Chlorophenyl)methanesulfonyl chloride, ≥97%, Thermo Scientific™

347.27 SEK - 730.85 SEK

Chemical Identifiers

CAS 77421-13-7
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.08
MDL Number MFCD04116369
InChI Key CHPZYFXSICSCNY-UHFFFAOYSA-N
Synonym 2-chlorophenyl methanesulfonyl chloride, 2-chloro-phenyl-methanesulfonyl chloride, chloro 2-chlorophenyl methyl sulfone, pubchem5500, acmc-1bfc0, 2-chlorobenzylsulfonyl chloride, benzenemethanesulfonylchloride, 2-chloro, 2-chloro-phenyl methanesulfonyl chloride, 2-chlorophenyl methane-sulfonyl chloride
PubChem CID 2757801
IUPAC Name (2-chlorophenyl)methanesulfonyl chloride
SMILES ClC1=CC=CC=C1CS(Cl)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10441504
View Documents
Thermo Scientific Maybridge
MO07829CB
250 mg Amber Glass Bottle
347.27 SEK
250mg
Please to purchase this item. Need a web account? Register with us today!
10536863
View Documents
Thermo Scientific Maybridge
MO07829DA
1 g Amber Glass Bottle
730.85 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

77421-13-7
225.08
CHPZYFXSICSCNY-UHFFFAOYSA-N
2757801
ClC1=CC=CC=C1CS(Cl)(=O)=O
C7H6Cl2O2S
MFCD04116369
2-chlorophenyl methanesulfonyl chloride, 2-chloro-phenyl-methanesulfonyl chloride, chloro 2-chlorophenyl methyl sulfone, pubchem5500, acmc-1bfc0, 2-chlorobenzylsulfonyl chloride, benzenemethanesulfonylchloride, 2-chloro, 2-chloro-phenyl methanesulfonyl chloride, 2-chlorophenyl methane-sulfonyl chloride
(2-chlorophenyl)methanesulfonyl chloride

Specifications

77421-13-7
97%
MFCD04116369
CHPZYFXSICSCNY-UHFFFAOYSA-N
(2-chlorophenyl)methanesulfonyl chloride
2757801
97%
97.0
C7H6Cl2O2S
2-chlorophenyl methanesulfonyl chloride, 2-chloro-phenyl-methanesulfonyl chloride, chloro 2-chlorophenyl methyl sulfone, pubchem5500, acmc-1bfc0, 2-chlorobenzylsulfonyl chloride, benzenemethanesulfonylchloride, 2-chloro, 2-chloro-phenyl methanesulfonyl chloride, 2-chlorophenyl methane-sulfonyl chloride
ClC1=CC=CC=C1CS(Cl)(=O)=O
225.08
225.09
(2-Chlorophenyl)methanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.