Learn More
Julabo™ 2-Collar Nuts
785.00 SEK - 1435.00 SEK
Specifications
Description | 2-Collar Nuts |
---|---|
For Use With (Equipment) | Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15214333
|
Julabo™
8970495 |
M24 x 1.5 Connections |
785.00 SEK
Pack of 2 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
15224333
|
Julabo™
8970496 |
M30 x 1.5 Connections |
1210.00 SEK
Pack of 2 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
15234333
|
Julabo™
8970497 |
M38 x 1.5 Connections |
1435.00 SEK
Pack of 2 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
Specifications
2-Collar Nuts | |
Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System |