missing translation for 'onlineSavingsMsg'
Learn More
Learn More
#2 Diesel Fuel Oil, 25% Weathered Fuel Standard, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Diesel Fuel | 68476-34-6 | 5000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 68476-34-6, 67-56-1 |
Packaging | 2mL amber prescored glass ampule |
CAS Min % | 0.5, 99.5 |
Quantity | 1 mL |
CAS Max % | 0.5, 99.5 |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Solvent or Matrix | Methanol-P&T |
Chemical Name or Material | #2 Diesel Fuel Oil, 25% Weathered |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'