missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-1-naphthoic acid, 98%, Thermo Scientific™

Product Code. 11405297
Click to view available options
Quantity:
25 g
100 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2224-00-2
Molecular Formula C13H11O3
Molecular Weight (g/mol) 215.23
MDL Number MFCD00004008
InChI Key MYFBSSDLYGWAHH-UHFFFAOYSA-M
Synonym 2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
PubChem CID 75210
IUPAC Name 2-ethoxynaphthalene-1-carboxylic acid
SMILES CCOC1=CC=C2C=CC=CC2=C1C([O-])=O

Specifications

Melting Point 143°C to 145°C
Quantity 100 g
Beilstein 2695612
Formula Weight 216.24
Percent Purity 98%
Chemical Name or Material 2-Ethoxy-1-naphthoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.