missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-3,5(6)-dimethylpyrazine, 99%, mixture of isomers, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Description
It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, chloroform and dichloromethane.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, chloroform and dichloromethane.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Spécification
Spécification
Density | 0.96 |
Boiling Point | 182°C |
Flash Point | 66°C (151°F) |
Refractive Index | 1.5015 |
Quantity | 25 g |
Solubility Information | Not miscible or difficult to mix in water. Soluble in alcohol,chloroform and dichloromethane. |
Formula Weight | 136.19 |
Percent Purity | 99% |
Chemical Name or Material | 2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'