Learn More
2-Ethylbutyl acetate, 97%, Thermo Scientific™
344.60 SEK - 917.00 SEK
Chemical Identifiers
CAS | 10031-87-5 |
---|---|
Molecular Formula | C8H16O2 |
Molecular Weight (g/mol) | 144.214 |
MDL Number | MFCD00009343 |
InChI Key | HQLKZWRSOHTERR-UHFFFAOYSA-N |
Synonym | acetic acid, 2-ethylbutyl ester, 2-ethyl butyl acetate, beta-ethylbutyl acetate, acetic acid 2-ethylbutyl ester, 1-butanol, 2-ethyl-, acetate, fema no. 2425, 2-ethylbutylacetat, 2-ethylbutyl=acetate, acmc-2097ot, 2-ethylbutyl acetateShow More |
PubChem CID | 61448 |
IUPAC Name | 2-ethylbutyl acetate |
SMILES | CCC(CC)COC(=O)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
10031-87-5 | |
144.214 | |
HQLKZWRSOHTERR-UHFFFAOYSA-N | |
61448 | |
CCC(CC)COC(=O)C |
C8H16O2 | |
MFCD00009343 | |
acetic acid, 2-ethylbutyl ester, 2-ethyl butyl acetate, beta-ethylbutyl acetate, acetic acid 2-ethylbutyl ester, 1-butanol, 2-ethyl-, acetate, fema no. 2425, 2-ethylbutylacetat, 2-ethylbutyl=acetate, acmc-2097ot, 2-ethylbutyl acetateShow More | |
2-ethylbutyl acetate |
Specifications
10031-87-5 | |
159°C to 160°C | |
C8H16O2 | |
MFCD00009343 | |
1746228 | |
HQLKZWRSOHTERR-UHFFFAOYSA-N | |
2-ethylbutyl acetate | |
61448 | |
97% |
0.87 | |
52°C (125°F) | |
1.41 | |
UN1177 | |
acetic acid, 2-ethylbutyl ester, 2-ethyl butyl acetate, beta-ethylbutyl acetate, acetic acid 2-ethylbutyl ester, 1-butanol, 2-ethyl-, acetate, fema no. 2425, 2-ethylbutylacetat, 2-ethylbutyl=acetate, acmc-2097ot, 2-ethylbutyl acetate | |
CCC(CC)COC(=O)C | |
144.214 | |
144.22 | |
2-Ethylbutyl acetate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 2-ETHYLBUTYL ACETATE
missing translation for 'einecsNumber' : 233-095-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.