missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals Product Code.: 15435236

2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15435236
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15435236

Brand: Thermo Scientific Alfa Aesar H59738.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1259022-70-2
Molecular Formula C15H20BFO4
Molecular Weight (g/mol) 294.13
MDL Number MFCD18427635
InChI Key ZFLOTGNTGSGUGD-UHFFFAOYSA-N
Synonym methyl 2-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-3-fluoro-4-4,4,5 5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate
PubChem CID 59627674
IUPAC Name methyl 2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
SMILES COC(=O)CC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Formula Weight 294.13
Percent Purity 96%
Chemical Name or Material 2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.