missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzaldehyde, 97%, Thermo Scientific Chemicals

329.00 SEK - 7320.00 SEK

Chemical Identifiers

CAS 446-52-6
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.114
MDL Number MFCD00003302
InChI Key ZWDVQMVZZYIAHO-UHFFFAOYSA-N
Synonym o-fluorobenzaldehyde, benzaldehyde, 2-fluoro, benzaldehyde, o-fluoro, unii-7zx20dxb9z, 2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, benzaldehyde, fluoro, ortho-fluorobenzaldehyde, 7zx20dxb9z, fluorobenzaldehyde
PubChem CID 67970
IUPAC Name 2-fluorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)F
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369619
SDS View Documents
Thermo Scientific Alfa Aesar
A13800.06
5 g
329.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11464874
SDS View Documents
Thermo Scientific Alfa Aesar
A13800.14
25 g
704.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
11474874
SDS View Documents
Thermo Scientific Alfa Aesar
A13800.22
100 g
1955.00 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
11484874
SDS View Documents
Thermo Scientific Alfa Aesar
A13800.36
500 g
7320.00 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2?-(2-Fluorobenzylidene)-2-hydroxybenzohydrazide was synthesized by the reaction of 2-hydroxybenzoylhydrazine with 2-fluorobenzaldehyde in ethanol. Synthesis of n-aryl indolines from 2-fluorobenzaldehyde dimethylhydrazone derivatives which is an approach to preparation of c(aryl)-n(amine) bond atropisomeric amines. The o-dialkylaminobenzaldehydes were conveniently prepared by nucleophilic displacement of the activated fluorine in 2-fluorobenzaldehyde with the required dialkylamine in hot dimethylformamide. N?-(2-Fluorobenzylidene)-2-(quinolin-8-yloxy) acetohydrazide methanol solvate in the reaction of quinolin-8-yloxyacetic acid hydrazide and 2-fluorobenzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-(2-Fluorobenzylidene)-2-hydroxybenzohydrazide was synthesized by the reaction of 2-hydroxybenzoylhydrazine with 2-fluorobenzaldehyde in ethanol. Synthesis of n-aryl indolines from 2-fluorobenzaldehyde dimethylhydrazone derivatives which is an approach to preparation of c(aryl)-n(amine) bond atropisomeric amines. The o-dialkylaminobenzaldehydes were conveniently prepared by nucleophilic displacement of the activated fluorine in 2-fluorobenzaldehyde with the required dialkylamine in hot dimethylformamide. N′-(2-Fluorobenzylidene)-2-(quinolin-8-yloxy) acetohydrazide methanol solvate in the reaction of quinolin-8-yloxyacetic acid hydrazide and 2-fluorobenzaldehyde.

Solubility
Insoluble in water.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, air, bases.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluorobenzaldehyde, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.