Learn More
2-Fluorophenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals
378.00 SEK - 3730.00 SEK
Chemical Identifiers
CAS | 2924-15-4 |
---|---|
Molecular Formula | C6H8ClFN2 |
Molecular Weight (g/mol) | 162.592 |
MDL Number | MFCD00012927 |
InChI Key | VFZYLSYYMHFPSY-UHFFFAOYSA-N |
Synonym | 2-fluorophenylhydrazine hydrochloride, 2-fluorophenyl hydrazine hydrochloride, 2-fluorophenylhydrazine hcl, 2-fluorophenylhydrazinehydrochloride, o-fluorophenylhydrazine hydrochloride, hydrazine, 2-fluorophenyl-, monohydrochloride, 1-2-fluorophenyl hydrazine hydrochloride, pubchem3347, c6h7fn2.hcl, acmc-209h7m |
PubChem CID | 2723910 |
IUPAC Name | (2-fluorophenyl)hydrazine;hydrochloride |
SMILES | C1=CC=C(C(=C1)NN)F.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404724
|
Thermo Scientific Alfa Aesar
A13716.03 |
1 g |
378.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414724
|
Thermo Scientific Alfa Aesar
A13716.06 |
5 g |
1135.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424724
|
Thermo Scientific Alfa Aesar
A13716.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2924-15-4 | |
162.592 | |
VFZYLSYYMHFPSY-UHFFFAOYSA-N | |
2723910 | |
C1=CC=C(C(=C1)NN)F.Cl |
C6H8ClFN2 | |
MFCD00012927 | |
2-fluorophenylhydrazine hydrochloride, 2-fluorophenyl hydrazine hydrochloride, 2-fluorophenylhydrazine hcl, 2-fluorophenylhydrazinehydrochloride, o-fluorophenylhydrazine hydrochloride, hydrazine, 2-fluorophenyl-, monohydrochloride, 1-2-fluorophenyl hydrazine hydrochloride, pubchem3347, c6h7fn2.hcl, acmc-209h7m | |
(2-fluorophenyl)hydrazine;hydrochloride |
Specifications
2924-15-4 | |
C6H8ClFN2 | |
3627985 | |
VFZYLSYYMHFPSY-UHFFFAOYSA-N | |
(2-fluorophenyl)hydrazine;hydrochloride | |
2723910 | |
98% |
∼205°C (decomposition) | |
MFCD00012927 | |
2-fluorophenylhydrazine hydrochloride, 2-fluorophenyl hydrazine hydrochloride, 2-fluorophenylhydrazine hcl, 2-fluorophenylhydrazinehydrochloride, o-fluorophenylhydrazine hydrochloride, hydrazine, 2-fluorophenyl-, monohydrochloride, 1-2-fluorophenyl hydrazine hydrochloride, pubchem3347, c6h7fn2.hcl, acmc-209h7m | |
C1=CC=C(C(=C1)NN)F.Cl | |
162.592 | |
162.6 | |
2-Fluorophenylhydrazine hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H315-H317-H319-H335-H351
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 220-885-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.