Learn More
2-(Fmoc-amino)-4-chlorobenzoic acid, 95%, Thermo Scientific™
715.00 SEK - 2982.11 SEK
Chemical Identifiers
CAS | 332121-92-3 |
---|---|
Molecular Formula | C22H16ClNO4 |
Molecular Weight (g/mol) | 393.823 |
MDL Number | MFCD02682157 |
InChI Key | BIPQSYSWHUYDEA-UHFFFAOYSA-N |
Synonym | 2-9h-fluoren-9-yl methoxy carbonyl amino-4-chlorobenzoic acid, 2-fmoc-amino-4-chlorobenzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxycarbonylamino benzoic acid, n-fmoc-4-chloroanthranilic acid, 2-n-fmoc-amino-4-chlorobenzoic acid, 4-chloro-2-fluoren-9-ylmethoxy carbonylamino benzoic acid, n-9-fluorenylmethoxycarbonyl-4-chloroanthranilic acid, 4-chloro-2-9h-fluoren-9-ylmethoxycarbonylamino-benzoic acid, benzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxy carbonyl amino |
PubChem CID | 7022993 |
IUPAC Name | 4-chloro-2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid |
SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=C(C=CC(=C4)Cl)C(=O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
332121-92-3 | |
393.823 | |
BIPQSYSWHUYDEA-UHFFFAOYSA-N | |
7022993 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=C(C=CC(=C4)Cl)C(=O)O |
C22H16ClNO4 | |
MFCD02682157 | |
2-9h-fluoren-9-yl methoxy carbonyl amino-4-chlorobenzoic acid, 2-fmoc-amino-4-chlorobenzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxycarbonylamino benzoic acid, n-fmoc-4-chloroanthranilic acid, 2-n-fmoc-amino-4-chlorobenzoic acid, 4-chloro-2-fluoren-9-ylmethoxy carbonylamino benzoic acid, n-9-fluorenylmethoxycarbonyl-4-chloroanthranilic acid, 4-chloro-2-9h-fluoren-9-ylmethoxycarbonylamino-benzoic acid, benzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxy carbonyl amino | |
4-chloro-2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid |
Specifications
332121-92-3 | |
C22H16ClNO4 | |
2-9h-fluoren-9-yl methoxy carbonyl amino-4-chlorobenzoic acid, 2-fmoc-amino-4-chlorobenzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxycarbonylamino benzoic acid, n-fmoc-4-chloroanthranilic acid, 2-n-fmoc-amino-4-chlorobenzoic acid, 4-chloro-2-fluoren-9-ylmethoxy carbonylamino benzoic acid, n-9-fluorenylmethoxycarbonyl-4-chloroanthranilic acid, 4-chloro-2-9h-fluoren-9-ylmethoxycarbonylamino-benzoic acid, benzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxy carbonyl amino | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=C(C=CC(=C4)Cl)C(=O)O | |
393.823 | |
393.83 | |
2-(Fmoc-amino)-4-chlorobenzoic acid |
218°C to 220°C (decomposition) | |
MFCD02682157 | |
BIPQSYSWHUYDEA-UHFFFAOYSA-N | |
4-chloro-2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid | |
7022993 | |
95% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.