missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals

172.97 SEK - 5372.53 SEK

Chemical Identifiers

CAS 818-61-1, 150-76-5
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002865
InChI Key OMIGHNLMNHATMP-UHFFFAOYSA-N
Synonym 2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
PubChem CID 13165
IUPAC Name 2-hydroxyethyl prop-2-enoate
SMILES C=CC(=O)OCCO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10786232
View Documents Promotion Details
Thermo Scientific Acros
334080050
5 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10368983
View Documents Promotion Details
Thermo Scientific
334082500
250 mL Glass bottle
981.02 SEK
250mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15750859
View Documents Promotion Details
Thermo Scientific Acros
334080010
1 L Glass bottle
2176.95 SEK
1 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10309090
View Documents Promotion Details
Thermo Scientific
334080025
2.5 L Glass bottle
5372.53 SEK
2.50 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

818-61-1, 150-76-5
116.12
OMIGHNLMNHATMP-UHFFFAOYSA-N
13165
C=CC(=O)OCCO
C5H8O3
MFCD00002865
2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
2-hydroxyethyl prop-2-enoate

Specifications

4mg KOH/g max.
96.0
-60°C
1.1060g/mL
99°C
96% min. (GC)
1.4490 to 1.451
1.106
Solubility in water: soluble in water
C=CC(=O)OCCO
200 to 450ppm MEHQ
13165
116.12
Oily Liquid
818-61-1, 150-76-5
100.0
Yellow to Brown
210°C to 215°C
Authentic
C5H8O3
MFCD00002865
2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl prop-2-enoate
116.12
9 mPa.s (20°C)
97%
2-Hydroxyethyl acrylate, Stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specif

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 212-454-9

Product Identifier
  • 2-Hydroxyethyl acrylate, stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only