missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-5-methylbenzonitrile, 97%, Thermo Scientific™
393.00 SEK - 1330.00 SEK
Chemical Identifiers
CAS | 53078-70-9 |
---|---|
Molecular Formula | C9H9NO |
Molecular Weight (g/mol) | 147.18 |
InChI Key | MYOASWSDRGHGLN-UHFFFAOYSA-N |
Synonym | 2-methoxy-5-methyl-benzonitrile, acmc-1an2t, benzonitrile,2-methoxy-5-methyl, 2-methoxy-5-methylbenzonitrile |
PubChem CID | 3801023 |
IUPAC Name | 2-methoxy-5-methylbenzonitrile |
SMILES | CC1=CC(=C(C=C1)OC)C#N |
Description
Chemical Identifiers
53078-70-9 | |
147.18 | |
2-methoxy-5-methyl-benzonitrile, acmc-1an2t, benzonitrile,2-methoxy-5-methyl, 2-methoxy-5-methylbenzonitrileShow More | |
2-methoxy-5-methylbenzonitrile |
C9H9NO | |
MYOASWSDRGHGLN-UHFFFAOYSA-N | |
3801023 | |
CC1=CC(=C(C=C1)OC)C#N |
Specifications
53078-70-9 | |
100.0 | |
90°C (1.0 mmHg) | |
Authentic | |
C9H9NO | |
MYOASWSDRGHGLN-UHFFFAOYSA-N | |
2-methoxy-5-methylbenzonitrile | |
3801023 | |
97% | |
2-Methoxy-5-methylbenzonitrile |
96.0 | |
Colorless to Yellow | |
>110°C | |
96% min. (GC) | |
2-methoxy-5-methyl-benzonitrile, acmc-1an2t, benzonitrile,2-methoxy-5-methyl, 2-methoxy-5-methylbenzonitrile | |
CC1=CC(=C(C=C1)OC)C#N | |
147.18 | |
147.18 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Methoxy-5-methylbenzonitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Methoxy-5-methylbenzonitrile, 97%, Thermo Scientific™