missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™

741.00 SEK - 2605.00 SEK

Chemical Identifiers

CAS 1240257-53-7
Molecular Formula C8H8BrF5OS
Molecular Weight (g/mol) 327.107
MDL Number MFCD16652488
InChI Key VOLXPNDUPQNACF-UHFFFAOYSA-N
Synonym 2-methoxy-5-pentafluorosulfur benzyl bromide, 2-bromomethyl-1-methoxy-4-pentafluoro-??-sulfanyl benzene, 2-methoxy-5-pentafluorothio benzyl bromide
PubChem CID 66523611
IUPAC Name [3-(bromomethyl)-4-methoxyphenyl]-pentafluoro-$l^{6}-sulfane
SMILES COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)CBr
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15458925
View Documents
Thermo Scientific Alfa Aesar
H34415.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15477357
View Documents
Thermo Scientific Alfa Aesar
H34415.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1240257-53-7
327.107
VOLXPNDUPQNACF-UHFFFAOYSA-N
66523611
COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)CBr
C8H8BrF5OS
MFCD16652488
2-methoxy-5-pentafluorosulfur benzyl bromide, 2-bromomethyl-1-methoxy-4-pentafluoro-??-sulfanyl benzene, 2-methoxy-5-pentafluorothio benzyl bromide
[3-(bromomethyl)-4-methoxyphenyl]-pentafluoro-$l^{6}-sulfane

Specifications

1240257-53-7
MFCD16652488
Moisture sensitive
VOLXPNDUPQNACF-UHFFFAOYSA-N
[3-(bromomethyl)-4-methoxyphenyl]-pentafluoro-$l^{6}-sulfane
66523611
97%
C8H8BrF5OS
UN3261
2-methoxy-5-pentafluorosulfur benzyl bromide, 2-bromomethyl-1-methoxy-4-pentafluoro-??-sulfanyl benzene, 2-methoxy-5-pentafluorothio benzyl bromide
COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)CBr
327.107
327.11
2-Methoxy-5-(pentafluorothio)benzyl bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.