missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-phenylthiazole, 97%, Thermo Scientific™

Product Code. 15498177
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1826-16-0
Molecular Formula C10H9NS
Molecular Weight (g/mol) 175.249
MDL Number MFCD00489998
InChI Key XROORURTAQOYLW-UHFFFAOYSA-N
Synonym 2-methyl-4-phenylthiazole, 2-methyl-4-phenyl-thiazole, thiazole, 2-methyl-4-phenyl, thiazole,2-methyl-4-phenyl, #
PubChem CID 584377
IUPAC Name 2-methyl-4-phenyl-1,3-thiazole
SMILES CC1=NC(=CS1)C2=CC=CC=C2

Specifications

Melting Point 51°C to 56°C
Quantity 5 g
Formula Weight 175.25
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-phenylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'