missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Methyl-5-nitroimidazole, 99%
Click to view available options
Quantity:
50 g
250 g
Description
Applications
2-Methyl-4(5)-nitroimidazole was used to study the mechanism of both the alkaline and acidic hydrolysis of tinidazole. It is mainly used to synthesize intermediate of medicines such as Metronidazole and dimetridazole.
Solubility
Slightly soluble in ethanol, easily soluble in diluted acid and diluted alkali.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
2-Methyl-4(5)-nitroimidazole was used to study the mechanism of both the alkaline and acidic hydrolysis of tinidazole. It is mainly used to synthesize intermediate of medicines such as Metronidazole and dimetridazole.
Solubility
Slightly soluble in ethanol, easily soluble in diluted acid and diluted alkali.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
Melting Point | 251°C to 255°C |
Density | 1.426g/mL |
Boiling Point | 399°C |
Quantity | 50 g |
Beilstein | 4032 |
Formula Weight | 127.1 |
Percent Purity | 99% |
Physical Form | Solid |
Chemical Name or Material | 2-Methyl-5-nitroimidazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'