Learn More
2-(Methylamino)-1H-purin-6(7H)-one, MedChemExpress
MedChemExpress 2-(Methylamino)-1H-purin-6(7H)-one (N2-Methylguanine) is a modified nucleoside. 2-(Methylamino)-1H-purin-6(7H)-one is an endogenous methylated nucleoside found in human fluids.
Brand: Medchem Express HY-101412
This item is not returnable.
View return policy
Description
2-(Methylamino)-1H-purin-6(7H)-one (N2-Methylguanine) is a modified nucleoside. 2-(Methylamino)-1H-purin-6(7H)-one is an endogenous methylated nucleoside found in human fluids.Specifications
2-(Methylamino)-1H-purin-6(7H)-one | |
10030-78-1 | |
50 mg | |
C6H7N5O | |
H2O : 1 mg/mL (6.06 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (6.06 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble) ∣THF : < 1 mg/mL (ultrasonic) (insoluble) ∣1M NaOH : < 1 mg/mL (ultrasonic;adjust pH to 12 with NaOH) (insoluble) ∣Acetone : < 1 mg/mL (ultrasonic) (insoluble) ∣0.1 M HCL : < 1 mg/mL (ultrasonic;adjust pH to 2 with HCl) (insoluble) ∣DMF : < 1 mg/mL (ultrasonic) (insoluble) | |
165.15 | |
98.0% |
Solid | |
Earth Yellow | |
Research | |
N2-methylguanine | |
O=C1N=C(NC)NC2=C1NC=N2 | |
165.15 | |
Research |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
2-(Methylamino)-1H-purin-6(7H)-one, MedChemExpress