missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)phenylboronic acid, 97%, Thermo Scientific™

301.10 SEK - 630.35 SEK

Chemical Identifiers

CAS 168618-42-6
Molecular Formula C7H9BO2S
Molecular Weight (g/mol) 168.02
MDL Number MFCD01318165
InChI Key QXBWTYBCNFKURT-UHFFFAOYSA-N
Synonym 2-methylthio phenylboronic acid, 2-methylthiophenylboronic acid, 2-methylthio phenyl boronic acid, 2-methylthio benzeneboronic acid, 2-thioanisoleboronic acid, 2-methylsulfanylphenyl boronic acid, 2-methylsulfanyl phenylboronic acid, 2-methylsulfanyl phenyl boranediol, 2-methylsulfanyl phenyl boronic acid, o-methylthio phenylboronic acid
PubChem CID 2773543
SMILES CSC1=CC=CC=C1B(O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10004587
SDS View Documents
Thermo Scientific Acros
393780010
1 g Glass Bottle
301.10 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10768278
SDS View Documents
Thermo Scientific Acros
393780050
5 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Methylthio)phenylboronic acid, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.