Learn More
2-Phenylpyridine, 98%, Thermo Scientific Chemicals
1080.00 SEK - 3360.00 SEK
Chemical Identifiers
CAS | 1008-89-5 |
---|---|
Molecular Formula | C11H9N |
Molecular Weight (g/mol) | 155.20 |
MDL Number | MFCD00006280,MFCD31699954 |
InChI Key | VQGHOUODWALEFC-UHFFFAOYSA-N |
Synonym | pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine???? |
PubChem CID | 13887 |
IUPAC Name | 2-phenylpyridine |
SMILES | C1=CC=C(C=C1)C1=CC=CC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392077
|
Thermo Scientific Alfa Aesar
L07861.09 |
10 g |
1080.00 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302087
|
Thermo Scientific Alfa Aesar
L07861.18 |
50 g |
3360.00 SEK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Phenylpyridine is used to produce 2-phenyl-pyridine-1-oxide. It is used in the manufacturing of organic light emitting diodes (OLEDs). It is used as an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Phenylpyridine is used to produce 2-phenyl-pyridine-1-oxide. It is used in the manufacturing of organic light emitting diodes (OLEDs). It is used as an intermediate.
Solubility
Fully miscible with ethanol, acetone, toluene, methylene chloride. Not miscible in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Chemical Identifiers
1008-89-5 | |
155.20 | |
VQGHOUODWALEFC-UHFFFAOYSA-N | |
13887 | |
C1=CC=C(C=C1)C1=CC=CC=N1 |
C11H9N | |
MFCD00006280,MFCD31699954 | |
pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine???? | |
2-phenylpyridine |
Specifications
1008-89-5 | |
1.086 | |
>110°C (230°F) | |
1.624 | |
110445 | |
Fully miscible with ethanol,acetone,toluene,methylene chloride. Not miscible in water. | |
C1=CC=C(C=C1)C1=CC=CC=N1 | |
155.20 | |
155.2 | |
2-Phenylpyridine |
-5°C | |
268°C to 270°C | |
C11H9N | |
MFCD00006280,MFCD31699954 | |
pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine???? | |
VQGHOUODWALEFC-UHFFFAOYSA-N | |
2-phenylpyridine | |
13887 | |
98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 213-763-1
missing translation for 'rtecsNumber' : UT7140900
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.