Learn More
2-Pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, ≥97%, Thermo Scientific™
566.29 SEK - 767.16 SEK
Chemical Identifiers
CAS | 116247-03-1 |
---|---|
Molecular Formula | C9H8N2O2S |
Molecular Weight (g/mol) | 208.24 |
MDL Number | MFCD02854711 |
InChI Key | LJGAQGZEJDQDAU-UHFFFAOYNA-N |
Synonym | 2-pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-3-pyridinyl, 2-pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid, 3-4-carboxy-4,5-dihydro-1,3-thiazol-2-yl pyridine, 2-3-pyridyl-1,3-thiazoline-4-carboxylic acid, acmc-20dw5p, 4-carboxy-4,5-dihydro-2-pyridin-3-yl-1,3-thiazole |
PubChem CID | 3130734 |
SMILES | OC(=O)C1CSC(=N1)C1=CN=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10422463
|
Thermo Scientific Maybridge
CC36901CB |
250 mg | Amber Glass Bottle |
566.29 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10391433
|
Thermo Scientific Maybridge
CC36901DA |
1 g | Amber Glass Bottle |
767.16 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
116247-03-1 | |
208.24 | |
LJGAQGZEJDQDAU-UHFFFAOYNA-N | |
3130734 |
C9H8N2O2S | |
MFCD02854711 | |
2-pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-3-pyridinyl, 2-pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid, 3-4-carboxy-4,5-dihydro-1,3-thiazol-2-yl pyridine, 2-3-pyridyl-1,3-thiazoline-4-carboxylic acid, acmc-20dw5p, 4-carboxy-4,5-dihydro-2-pyridin-3-yl-1,3-thiazole | |
OC(=O)C1CSC(=N1)C1=CN=CC=C1 |
Specifications
116247-03-1 | |
100.0 | |
C9H8N2O2S | |
2-pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-3-pyridinyl, 2-pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid, 3-4-carboxy-4,5-dihydro-1,3-thiazol-2-yl pyridine, 2-3-pyridyl-1,3-thiazoline-4-carboxylic acid, acmc-20dw5p, 4-carboxy-4,5-dihydro-2-pyridin-3-yl-1,3-thiazole | |
OC(=O)C1CSC(=N1)C1=CN=CC=C1 | |
208.24 | |
208.24 | |
2-Pyrid-3-yl-4, 5-dihydro-1, 3-thiazole-4-carboxylic acid |
96.5 | |
97% | |
MFCD02854711 | |
LJGAQGZEJDQDAU-UHFFFAOYNA-N | |
2-(pyridin-3-yl)-4,5-dihydro-1,3-thiazole-4-carboxylic acid | |
3130734 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.