missing translation for 'onlineSavingsMsg'
Learn More
2-(tert-Butyl)-4,6-dimethylphenol, 97%, Thermo Scientific Chemicals Product Code.: 15760919
compliance-icons compliance-icons compliance-icons

2-(tert-Butyl)-4,6-dimethylphenol, 97%, Thermo Scientific Chemicals

Product Code. 15760919
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 15760919

Brand: Thermo Scientific Acros 462570500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1879-09-0
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00002234
InChI Key OPLCSTZDXXUYDU-UHFFFAOYSA-N
Synonym 2,4-dimethyl-6-tert-butylphenol, 2-tert-butyl-4,6-dimethylphenol, 6-tert-butyl-2,4-dimethylphenol, topanol a, 6-tert-butyl-2,4-xylenol, 6-t-butyl-2,4-xylenol, prodox 340, phenol, 2-1,1-dimethylethyl-4,6-dimethyl, 6-t-butyl-2,4-dimethylphenol, 2,4-xylenol, 6-tert-butyl
PubChem CID 15884
IUPAC Name 2-tert-butyl-4,6-dimethylphenol
SMILES CC1=CC(C)=C(O)C(=C1)C(C)(C)C

Specifications

Melting Point 22.0°C to 23.0°C
Boiling Point 249.0°C
Quantity 50 g
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2(OH)C6H2C(CH3)3
Formula Weight 178.27
Percent Purity 97%
Chemical Name or Material 2-(tert-Butyl)-4,6-dimethylphenol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443886 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number462570500,462570100
Lot NumberA0443886 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog Number462570500,462570100
Lot NumberA0413285 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number462570500,462570100
Lot NumberA0412471 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number462570500,462570100
Lot NumberA0410211 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number462570500,462570100
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-(tert-Butyl)-4,6-dimethylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : 2,4-Dimethyl-6-tert-butylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(tert-Butyl)-4,6-dimethylphenol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.