missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals

392.00 SEK - 1360.00 SEK

Chemical Identifiers

CAS 1743-86-8
Molecular Formula C8H4F3NS
Molecular Weight (g/mol) 203.182
MDL Number MFCD00039644
InChI Key FCEKLQPJGXIQRY-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester
PubChem CID 737162
IUPAC Name 1-isothiocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352876
SDS View Documents
Thermo Scientific Alfa Aesar
L10586.03
1 g
392.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11362876
SDS View Documents
Thermo Scientific Alfa Aesar
L10586.06
5 g
1360.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.