Learn More
2-(Trimethylsiloxy)ethyl methacrylate, 94%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Product Code. 11369315
Click to view available options
Quantity:
10 g
50 g
2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17407-09-9
Molecular Formula C9H18O3Si
Molecular Weight (g/mol) 202.33
MDL Number MFCD00053869
InChI Key WUGOQZFPNUYUOO-UHFFFAOYSA-N
Synonym 2-trimethylsilyl oxy ethyl methacrylate, 2-trimethylsilyloxy ethyl methacrylate, methacryloxyethoxytrimethylsilane, 2-trimethylsiloxy ethyl methacrylate, trimethylsiloxy-ethylmethacrylate, acmc-1buxw, trimethylsilyloxyethyl-methacrylate, 2-trimethylsiloxyethyl methacrylate, 2-trimethylsiloxy ethylmethacrylate
PubChem CID 87094
IUPAC Name 2-trimethylsilyloxyethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCO[Si](C)(C)C
Specifications
Density 0.995
Boiling Point 65°C (0.9 mmHg)
Flash Point 42°C (107°F)
Refractive Index 1.43
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Difficult to mix.
Formula Weight 202.33
Percent Purity 94%
Chemical Name or Material 2-(Trimethylsiloxy)ethyl methacrylate, stabilized with 100ppm 4-methoxyphenol