Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Brand: RETSCH 60.123.006300
2151.75 SEK valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Specifications
1Each | |
Test Sieve | |
Stainless Steel | |
25 mm | |
200 mm | |
Stainless Steel |
6.3 mm | |
ASTM E 11 | |
Shaker AS 300 | |
For separation, fractioning, particle size determination | |
Stainless Steel |