missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Pyridinio)-1-propanesulfonate, 99%, Thermo Scientific Chemicals
634.60 SEK - 2913.37 SEK
Chemical Identifiers
CAS | 15471-17-7 |
---|---|
Molecular Formula | C8H11NO3S |
Molecular Weight (g/mol) | 201.24 |
MDL Number | MFCD00064468 |
InChI Key | REEBJQTUIJTGAL-UHFFFAOYSA-N |
Synonym | 3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-iumShow More |
PubChem CID | 84929 |
IUPAC Name | 3-pyridin-1-ium-1-ylpropane-1-sulfonate |
SMILES | [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15547039
|
Thermo Scientific Acros
459670500 |
50 g |
634.60 SEK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15557039
|
Thermo Scientific Acros
459672500 |
250 g |
2913.37 SEK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15471-17-7 | |
201.24 | |
REEBJQTUIJTGAL-UHFFFAOYSA-N | |
84929 | |
[O-]S(=O)(=O)CCC[N+]1=CC=CC=C1 |
C8H11NO3S | |
MFCD00064468 | |
3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-iumShow More | |
3-pyridin-1-ium-1-ylpropane-1-sulfonate |
Specifications
15471-17-7 | |
1% max. | |
98.5% min. (HPLC) | |
MFCD00064468 | |
(10% in water) Clear solution | |
[O-]S(=O)(=O)CCC[N+]1=CC=CC=C1 | |
201.24 | |
201.25 | |
3-(1-Pyridinio)-1-propanesulfonate |
275.0°C | |
Authentic | |
C8H11NO3S | |
3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium | |
REEBJQTUIJTGAL-UHFFFAOYSA-N | |
3-pyridin-1-ium-1-ylpropane-1-sulfonate | |
84929 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 239-491-3
Product Identifier
- 3-(1-Pyridinio)-1-propanesulfonate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-(1-Pyridinio)-1-propanesulfonate, 99%, Thermo Scientific Chemicals