Learn More
3-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals
1435.00 SEK - 4595.00 SEK
Chemical Identifiers
CAS | 20173-24-4 |
---|---|
Molecular Formula | C7H11N2 |
Molecular Weight (g/mol) | 123.18 |
MDL Number | MFCD00191604 |
InChI Key | NAHHNSMHYCLMON-UHFFFAOYSA-O |
Synonym | 3-2-aminoethyl pyridine, 2-pyridin-3-yl ethanamine, 3-aminoethylpyridine, 3-pyridineethanamine, 2-pyridin-3-yl ethan-1-amine, 2-3-pyridyl ethylamine, 3-pyridineethaneamine, 3-pyridine ethanamine, 2-pyridin-3-yl-ethylamine, 2-3-pyridinyl ethanamine |
PubChem CID | 854051 |
SMILES | [NH3+]CCC1=CC=CN=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392977
|
Thermo Scientific Alfa Aesar
L16311.03 |
1 g |
1435.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302987
|
Thermo Scientific Alfa Aesar
L16311.06 |
5 g |
4595.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
Chemical Identifiers
20173-24-4 | |
123.18 | |
NAHHNSMHYCLMON-UHFFFAOYSA-O | |
854051 |
C7H11N2 | |
MFCD00191604 | |
3-2-aminoethyl pyridine, 2-pyridin-3-yl ethanamine, 3-aminoethylpyridine, 3-pyridineethanamine, 2-pyridin-3-yl ethan-1-amine, 2-3-pyridyl ethylamine, 3-pyridineethaneamine, 3-pyridine ethanamine, 2-pyridin-3-yl-ethylamine, 2-3-pyridinyl ethanamine | |
[NH3+]CCC1=CC=CN=C1 |
Specifications
20173-24-4 | |
Amine-like | |
1.5518 | |
UN2735 | |
Air Sensitive | |
Slightly soluble in water. | |
[NH3+]CCC1=CC=CN=C1 | |
123.18 | |
122.17 | |
3-(2-Aminoethyl)pyridine |
119°C to 120°C (19 mmHg) | |
C7H11N2 | |
MFCD00191604 | |
111288 | |
3-2-aminoethyl pyridine, 2-pyridin-3-yl ethanamine, 3-aminoethylpyridine, 3-pyridineethanamine, 2-pyridin-3-yl ethan-1-amine, 2-3-pyridyl ethylamine, 3-pyridineethaneamine, 3-pyridine ethanamine, 2-pyridin-3-yl-ethylamine, 2-3-pyridinyl ethanamine | |
NAHHNSMHYCLMON-UHFFFAOYSA-O | |
2-(pyridin-3-yl)ethan-1-aminium | |
854051 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.