missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluoropyrrolidine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11379735
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163457-23-6
Molecular Formula C4H8ClF2N
Molecular Weight (g/mol) 143.562
MDL Number MFCD03788948
InChI Key YYVPZQADFREIFR-UHFFFAOYSA-N
Synonym 3,3-difluoropyrrolidine hydrochloride, 3,3-difluoropyrrolidine hcl, 3,3-difluoropyrrolidinehydrochloride, pyrrolidine, 3,3-difluoro-, hydrochloride, pyrrolidine, 3,3-difluoro-, hydrochloride 1:1, 3,3-difluoro-pyrrolidine hcl, 3,3-difluoropyrrolidine hcl salt, 3,3-difluoropyrrolidine, chloride, 3,3-difluoro-pyrrolidinehydrochloride, pubchem9358
PubChem CID 24903482
IUPAC Name 3,3-difluoropyrrolidine;hydrochloride
SMILES C1CNCC1(F)F.Cl

Specifications

Melting Point 130°C to 134°C
Quantity 100 mg
Sensitivity Hygroscopic
Formula Weight 143.56
Percent Purity 98%
Chemical Name or Material 3,3-Difluoropyrrolidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.