Learn More
3-[3-(Dimethylamino)propoxy]-N-methylbenzylamine, 97%, Thermo Scientific™
650.00 SEK - 1565.00 SEK
Chemical Identifiers
CAS | 910037-03-5 |
---|---|
Molecular Formula | C13H22N2O |
Molecular Weight (g/mol) | 222.33 |
MDL Number | MFCD09064992 |
InChI Key | ODWHICGCLWFLBT-UHFFFAOYSA-N |
Synonym | 3-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-3-methylamino methyl phenoxy propyl amine, 3-3-dimethylamino propoxy phenyl methyl methyl amine, n,n-dimethyl-3-3-methylamino methyl phenoxy propan-1-amine, benzenemethanamine, 3-3-dimethylamino propoxy-n-methyl, benzenemethanamine,3-3-dimethylamino propoxy-n-methyl |
PubChem CID | 24229606 |
IUPAC Name | N,N-dimethyl-3-[3-(methylaminomethyl)phenoxy]propan-1-amine |
SMILES | CNCC1=CC(OCCCN(C)C)=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10245484
|
Thermo Scientific Maybridge
CC59446CB |
250 mg | Amber Glass Bottle |
650.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10205534
|
Thermo Scientific Maybridge
CC59446DA |
1 g | Amber Glass Bottle |
1565.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
910037-03-5 | |
222.33 | |
ODWHICGCLWFLBT-UHFFFAOYSA-N | |
24229606 | |
CNCC1=CC(OCCCN(C)C)=CC=C1 |
C13H22N2O | |
MFCD09064992 | |
3-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-3-methylamino methyl phenoxy propyl amine, 3-3-dimethylamino propoxy phenyl methyl methyl amine, n,n-dimethyl-3-3-methylamino methyl phenoxy propan-1-amine, benzenemethanamine, 3-3-dimethylamino propoxy-n-methyl, benzenemethanamine,3-3-dimethylamino propoxy-n-methyl | |
N,N-dimethyl-3-[3-(methylaminomethyl)phenoxy]propan-1-amine |
Specifications
910037-03-5 | |
100.0 | |
C13H22N2O | |
3-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-3-methylamino methyl phenoxy propyl amine, 3-3-dimethylamino propoxy phenyl methyl methyl amine, n,n-dimethyl-3-3-methylamino methyl phenoxy propan-1-amine, benzenemethanamine, 3-3-dimethylamino propoxy-n-methyl, benzenemethanamine,3-3-dimethylamino propoxy-n-methyl | |
CNCC1=CC(OCCCN(C)C)=CC=C1 | |
222.33 | |
222.33 | |
3-[3-(Dimethylamino)propoxy]-N-methylbenzylamine |
94.5 | |
95% | |
MFCD09064992 | |
ODWHICGCLWFLBT-UHFFFAOYSA-N | |
N,N-dimethyl-3-[3-(methylaminomethyl)phenoxy]propan-1-amine | |
24229606 | |
95% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger