Learn More
3-(4-Biphenyl)propionic acid, 98%, Thermo Scientific Chemicals Product Code.: 15405487
3-(4-Biphenyl)propionic acid, 98%, Thermo Scientific Chemicals
Product Code. 15405487
1 g
Click to view available options
Quantity:
1 g
5 g
Product Code. 15405487
Brand: Thermo Scientific Alfa Aesar H32729.03
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 35888-99-4
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD06823979
InChI Key MVFHRQWYCXYYMU-UHFFFAOYSA-N
Synonym 2 4-biphenyl propionic acid, 3-4-phenylphenyl propanoic acid, 3-4-biphenyl propionic acid, 3-1,1'-biphenyl-4-yl propanoic acid, 3-biphenyl-4-yl propanoic acid, 1,1'-biphenyl-4-propanoic acid, 2 para-biphenyl propionic acid, biphenyl-4-propanoic acidShow MoreShow Less
PubChem CID 3082434
IUPAC Name 3-(4-phenylphenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)C1=CC=CC=C1
Specifications
Melting Point 149°C to 152°C
Quantity 1 g
Formula Weight 226.28
Percent Purity 98%
Chemical Name or Material 3-(4-Biphenyl)propionic acid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10173086 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32729.03,H32729.06
compliance-icons
Product Identifier
  • 3-(4-Biphenyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : [1,1-Biphenyl]-4-propanoic acid
RUO – Research Use Only
Product Content Correction
Product Title
3-(4-Biphenyl)propionic acid, 98%, Thermo Scientific Chemicals >