missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine, ≥97%, Thermo Scientific™

383.58 SEK - 2088.13 SEK

Chemical Identifiers

CAS 126417-82-1
Molecular Formula C10H10ClN3
Molecular Weight (g/mol) 207.661
MDL Number MFCD00084872
InChI Key ZSFCXTGPGSAWFT-UHFFFAOYSA-N
Synonym 3-4-chlorophenyl-1-methyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-2-methylpyrazol-3-amine, 1h-pyrazol-5-amine,3-4-chlorophenyl-1-methyl, 5-amino-3-4-chlorophenyl-1-methyl-1h-pyrazole, 3-4-chlorophenyl-1-methyl-1h-pyrazol-5-ylamine, 5-4-chloro-phenyl-2-methyl-2h-pyrazol-3-ylamine, maybridge1_004889, acmc-1c52v, 5-4-chlorophenyl-2-methyl-3-pyrazolamine, 5-4-chlorophenyl-2-methyl-pyrazol-3-amine
PubChem CID 2735304
IUPAC Name 5-(4-chlorophenyl)-2-methylpyrazol-3-amine
SMILES CN1C(=CC(=N1)C2=CC=C(C=C2)Cl)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10575063
SDS View Documents
Thermo Scientific Maybridge
KM04038DA
1 g
383.58 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10432393
SDS View Documents
Thermo Scientific Maybridge
KM04038EA
10 g
2088.13 SEK
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

126417-82-1
207.661
ZSFCXTGPGSAWFT-UHFFFAOYSA-N
2735304
CN1C(=CC(=N1)C2=CC=C(C=C2)Cl)N
C10H10ClN3
MFCD00084872
3-4-chlorophenyl-1-methyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-2-methylpyrazol-3-amine, 1h-pyrazol-5-amine,3-4-chlorophenyl-1-methyl, 5-amino-3-4-chlorophenyl-1-methyl-1h-pyrazole, 3-4-chlorophenyl-1-methyl-1h-pyrazol-5-ylamine, 5-4-chloro-phenyl-2-methyl-2h-pyrazol-3-ylamine, maybridge1_004889, acmc-1c52v, 5-4-chlorophenyl-2-methyl-3-pyrazolamine, 5-4-chlorophenyl-2-methyl-pyrazol-3-amine
5-(4-chlorophenyl)-2-methylpyrazol-3-amine

Specifications

126417-82-1
Amber glass bottle
C10H10ClN3
3-4-chlorophenyl-1-methyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-2-methylpyrazol-3-amine, 1h-pyrazol-5-amine,3-4-chlorophenyl-1-methyl, 5-amino-3-4-chlorophenyl-1-methyl-1h-pyrazole, 3-4-chlorophenyl-1-methyl-1h-pyrazol-5-ylamine, 5-4-chloro-phenyl-2-methyl-2h-pyrazol-3-ylamine, maybridge1_004889, acmc-1c52v, 5-4-chlorophenyl-2-methyl-3-pyrazolamine, 5-4-chlorophenyl-2-methyl-pyrazol-3-amine
CN1C(=CC(=N1)C2=CC=C(C=C2)Cl)N
207.661
207.66
3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
97.0
97%
MFCD00084872
ZSFCXTGPGSAWFT-UHFFFAOYSA-N
5-(4-chlorophenyl)-2-methylpyrazol-3-amine
2735304
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.