missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals Product Code.: 11408346

3,4-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11408346
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11408346

Brand: Thermo Scientific Alfa Aesar A15388.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

Solubility
Soluble in ethanol, ether, toluene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5973-71-7
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00016612
InChI Key POQJHLBMLVTHAU-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehydeShow MoreShow Less
PubChem CID 22278
IUPAC Name 3,4-dimethylbenzaldehyde
SMILES CC1=C(C=C(C=C1)C=O)C

Specifications

Density 1.017
Boiling Point 226°C
Odor Almond-like
Refractive Index 1.551
Quantity 25 g
Beilstein 1634270
Sensitivity Air Sensitive
Solubility Information Soluble in ethanol,ether,toluene. Insoluble in water.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 3,4-Dimethylbenzaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10149396 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberA15388.14,A15388.06
Lot Number10058000 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15388.14,A15388.06
Lot NumberF7594A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15388.14,A15388.06
Lot NumberF8224A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15388.14,A15388.06
Lot Number10149396 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15388.14,A15388.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.