missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dimethylphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 11449953

3,4-Dimethylphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11449953
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11449953

Brand: Thermo Scientific Alfa Aesar A13027.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60481-51-8
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00052270
InChI Key YYMIOVAEQIEPET-UHFFFAOYSA-N
Synonym 3,4-dimethylphenylhydrazine hydrochloride, 3,4-dimethylphenyl hydrazine hydrochloride, 1-3,4-dimethylphenyl hydrazine hydrochloride, hydrazine, 3,4-dimethylphenyl-, monohydrochloride, 4-hydrazino-o-xylene hydrochloride, 3,4-dimethylphenyl hydrazine hydrochloride 1:x, hydrazine, 3,4-dimethylphenyl-, hydrochloride, 3,4-dimethylphenyl diazane hydrochloride, pubchem7555, 3,4-dimethylphenylhydrazinehydrochloride
PubChem CID 173740
IUPAC Name (3,4-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C=C(C=C1)NN)C.Cl

Specifications

Melting Point ∼195°C (decomposition)
Quantity 50 g
Beilstein 6096060
Solubility Information Soluble in water.
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylphenylhydrazine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberF2069B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13027.30,A13027.18,A13027.09
Lot Number10099143 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13027.30,A13027.18,A13027.09
Lot NumberH8477A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13027.30,A13027.18,A13027.09
Lot Number10192374 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13027.30,A13027.18,A13027.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dimethylphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.