Learn More
3,4-Dimethylthiophenol, 98%, Thermo Scientific Chemicals
669.00 SEK - 5995.00 SEK
Chemical Identifiers
CAS | 18800-53-8 |
---|---|
Molecular Formula | C8H10S |
Molecular Weight (g/mol) | 138.228 |
MDL Number | MFCD00010023 |
InChI Key | IDKCKPBAFOIONK-UHFFFAOYSA-N |
Synonym | 3,4-dimethylthiophenol, benzenethiol, 3,4-dimethyl, 3,4-dimethyl thiophenol, o-xylene-4-thiol, 4-mercapto-o-xylene, 3,4-dimethylbenzene-1-thiol, pubchem6812, 3,4-dimethyl-thiophenol, acmc-1bu1r, 3,4-dimethyl-benzenethiol |
PubChem CID | 87799 |
IUPAC Name | 3,4-dimethylbenzenethiol |
SMILES | CC1=C(C=C(C=C1)S)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420757
|
Thermo Scientific Alfa Aesar
B23168.03 |
1 g |
669.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430757
|
Thermo Scientific Alfa Aesar
B23168.06 |
5 g |
1940.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440757
|
Thermo Scientific Alfa Aesar
B23168.14 |
25 g |
5995.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18800-53-8 | |
138.228 | |
IDKCKPBAFOIONK-UHFFFAOYSA-N | |
87799 | |
CC1=C(C=C(C=C1)S)C |
C8H10S | |
MFCD00010023 | |
3,4-dimethylthiophenol, benzenethiol, 3,4-dimethyl, 3,4-dimethyl thiophenol, o-xylene-4-thiol, 4-mercapto-o-xylene, 3,4-dimethylbenzene-1-thiol, pubchem6812, 3,4-dimethyl-thiophenol, acmc-1bu1r, 3,4-dimethyl-benzenethiol | |
3,4-dimethylbenzenethiol |
Specifications
18800-53-8 | |
218°C | |
Unpleasant | |
1.5736 | |
UN2810 | |
Air Sensitive | |
IDKCKPBAFOIONK-UHFFFAOYSA-N | |
3,4-dimethylbenzenethiol | |
87799 | |
98% |
1.027 | |
90°C (194°F) | |
C8H10S | |
MFCD00010023 | |
2040353 | |
3,4-dimethylthiophenol, benzenethiol, 3,4-dimethyl, 3,4-dimethyl thiophenol, o-xylene-4-thiol, 4-mercapto-o-xylene, 3,4-dimethylbenzene-1-thiol, pubchem6812, 3,4-dimethyl-thiophenol, acmc-1bu1r, 3,4-dimethyl-benzenethiol | |
CC1=C(C=C(C=C1)S)C | |
138.228 | |
138.23 | |
3,4-Dimethylthiophenol |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 242-587-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only