missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)thiobenzamide, 97%, Thermo Scientific™

Product Code. 15448307
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15884-65-8
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD02677739
InChI Key YHXXBQMLJHUUJU-UHFFFAOYSA-N
Synonym 2h-1,3-benzodioxole-5-carbothioamide, thiopiperonylamide, benzo d 1,3 dioxole-5-carbothioamide, 3,4-methylenedioxy thiobenzamide, benzo 1,3 dioxole-5-carbothioic acid amide, 1,3-benzodioxole-5-thiocarboxamide, 3,4-methylenedioxy benz-thioamide, 2h-benzo 3,4-d 1,3-dioxolen-5-ylaminomethane-1-thione
PubChem CID 2776126
IUPAC Name 1,3-benzodioxole-5-carbothioamide
SMILES C1OC2=C(O1)C=C(C=C2)C(=S)N

Specifications

Quantity 250 mg
Formula Weight 181.21
Percent Purity 97%
Chemical Name or Material 3,4-(Methylenedioxy)thiobenzamide
compliance-icons
Product Identifier
  • 3,4-(Methylenedioxy)thiobenzamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Benzodioxole-5-carbothioamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.