missing translation for 'onlineSavingsMsg'
Learn More

3,5,5-Trimethylhexanoic acid, 97%, Thermo Scientific Chemicals

518.00 SEK - 1740.00 SEK

Chemical Identifiers

CAS 3302-10-1
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00020507
InChI Key OILUAKBAMVLXGF-UHFFFAOYNA-N
Synonym hexanoic acid, 3,5,5-trimethyl, 3,5,5-trimethyl-hexanoic acid, 3,5,5-trimethylhexanoicacid, unii-aw943q219o, acmc-1assi, dsstox_cid_9254, 3,5-trimethylhexanoic acid, dsstox_rid_78735, dsstox_gsid_29254, 3,5,5 trimethylhexanoic acid
PubChem CID 90960
IUPAC Name 3,5,5-trimethylhexanoic acid
SMILES CC(CC(=O)O)CC(C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318076
SDS View Documents
Thermo Scientific Alfa Aesar
L13360.AE
100 mL
518.00 SEK
100mL
Please to purchase this item. Need a web account? Register with us today!
11328076
SDS View Documents
Thermo Scientific Alfa Aesar
L13360.AP
500 mL
1740.00 SEK
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3,5,5-Trimethylhexanoic acid, is an important organic intermediate. It is also used as an an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5,5-Trimethylhexanoic acid, is an important organic intermediate. It is also used as an an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Miscible with the usual organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5,5-Trimethylhexanoic acid, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.