missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11412918
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421-49-4
Molecular Formula C30H42NiO6
Molecular Weight (g/mol) 557.35
MDL Number MFCD00008827
InChI Key VIUVLVWUWLHYGT-UHFFFAOYSA-L
Synonym 3,5-di-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-9f0i7yag34, 3,5-di-t-butyl-4-hydroxybenzoic acid, 3,5-bis-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-di-tert-butyl-4-hydroxy, 3,5-di tert-butyl-4-hydroxybenzoic acid, 3,5-di-tert-butyl-4-hydroxy benzoic acid, 3,5-bis tert-butyl-4-hydroxybenzoic acid, pubchem15681
PubChem CID 15007
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzoic acid
SMILES [Ni++].CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O.CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O

Specifications

Melting Point 206°C to 209°C
Quantity 50 g
Beilstein 2216948
Solubility Information Insoluble in water.
Formula Weight 250.34
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'