missing translation for 'onlineSavingsMsg'
Learn More
3,5-Difluoroaniline, 98+%, Thermo Scientific Chemicals Product Code.: 11399519

3,5-Difluoroaniline, 98+%, Thermo Scientific Chemicals

Product Code. 11399519
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11399519

Brand: Thermo Scientific Alfa Aesar A16528.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-39-4
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007763
InChI Key KQOIBXZRCYFZSO-UHFFFAOYSA-N
Synonym benzenamine, 3,5-difluoro, 3,5-difluorobenzenamine, 3,5-difluorophenylamine, 5an, 3,5-difluoranilin, 3,5-difluoroanilin, 3,5 difluoroaniline, 3.5-difluoroaniline, pubchem2296, 3, 5-difluoroaniline
PubChem CID 96595
IUPAC Name 3,5-difluoroaniline
SMILES C1=C(C=C(C=C1F)F)N

Specifications

Melting Point 37°C to 41°C
Density 1.295
Boiling Point 80°C (20 mmHg)
Flash Point 75°C (167°F)
Odor Characteristic
Quantity 100 g
UN Number UN2811
Beilstein 2802286
Formula Weight 129.11
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluoroaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Difluoroaniline, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.