missing translation for 'onlineSavingsMsg'
Learn More
3,5-Difluorobenzyl chloride, 98%, Thermo Scientific™ Product Code.: 11325455

3,5-Difluorobenzyl chloride, 98%, Thermo Scientific™

Product Code. 11325455
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11325455

Brand: Thermo Scientific Alfa Aesar L19646.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220141-71-9
Molecular Formula C7H5ClF2
Molecular Weight (g/mol) 162.56
MDL Number MFCD01090990
InChI Key VNGSMSFVLAAOGK-UHFFFAOYSA-N
Synonym 3,5-difluorobenzyl chloride, 1-chloromethyl-3,5-difluorobenzene, 3,5-difluorobenzylchloride, alpha-chloro-3,5-difluorotoluene, benzene, 1-chloromethyl-3,5-difluoro, 1-chloromethyl-3,5-difluoro-benzene, benzene,1-chloromethyl-3,5-difluoro, acmc-1cb2p, ksc493k5f
PubChem CID 2736955
IUPAC Name 1-(chloromethyl)-3,5-difluorobenzene
SMILES FC1=CC(CCl)=CC(F)=C1

Specifications

Boiling Point 82°C (22 mmHg)
Refractive Index 1.4885
Quantity 1 g
UN Number UN3265
Beilstein 7473788
Formula Weight 162.57
Percent Purity 98%
Chemical Name or Material 3,5-Difluorobenzyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Difluorobenzyl chloride, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.