missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester, 95%, Thermo Scientific™ Product Code.: 15435145

3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15435145
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15435145

Brand: Thermo Scientific Alfa Aesar H32478.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126779-11-0
Molecular Formula C16H27BN2O3
Molecular Weight (g/mol) 306.213
MDL Number MFCD12031452
InChI Key ZNDYJQBCSOGOJQ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1-tetrahydro-2h-pyran-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-thp-3,5-dimethylpyrazole-4-boronic acid pinacol ester, 3,5-dimethyl-1-2-tetrahydropyranyl-1h-pyrazole-4-boronic acid pinacol ester, 3,5-dimethyl-1-oxan-2-yl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 3,5-dimethyl-1-oxan-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-thp-3,5-dimethylpyrazole-4-boronic acid, pinacol ester, 3,5-dimethyl-1-oxan-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 3,5-dimethyl-1-tetrahydro-pyran-2-yl-1h-pyrazoleboronic acid pinacol ester, 3,5-dimethyl-1-tetrahydro-2h-pyran-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazolShow MoreShow Less
PubChem CID 53217144
IUPAC Name 3,5-dimethyl-1-(oxan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N(N=C2C)C3CCCCO3)C

Specifications

Quantity 1 g
Formula Weight 306.22
Percent Purity 95%
Chemical Name or Material 3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSP417-2 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.